The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114121.2 | NP_001107593.1 | 3 - 4 | 1185 | 80 |
NM_001114122.2 | NP_001107594.1 | 3 - 4 | 1185 | 80 | |
NM_001244846.1 | NP_001231775.1 | 3 - 4 | 1185 | 80 | |
NM_001274.5 | NP_001265.2 | 3 - 4 | 528 | 80 | |
NR_045205.1 | 3 - 4 | 722 | 80 | ||
XM_017017146.1 | XP_016872635.1 | 3 - 4 | 372 | 80 | |
GenBank mRNA | AB451222.1 | - | 2 - 3 | 288 | 80 |
AB451345.1 | - | 2 - 3 | 288 | 80 | |
AF016582.1 | - | 2 - 3 | 322 | 80 | |
AF032874.1 | - | 2 - 3 | 288 | 80 | |
AK292549.1 | - | 3 - 4 | 722 | 80 | |
AK293143.1 | - | 3 - 4 | 350 | 80 | |
BC004202.2 | - | 3 - 4 | 347 | 80 | |
BC017575.1 | - | 3 - 4 | 446 | 80 | |
BP207812.1 | - | 4 - 5 | 490 | 80 | |
DA567616.1 | - | 3 - 4 | 419 | 80 |