The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014046.3 | NP_054765.1 | 3 - 4 | 435 | 86 |
GenBank mRNA | AF100761.1 | - | 3 - 4 | 290 | 86 |
AF151017.1 | - | 3 - 4 | 316 | 86 | |
AK294086.1 | - | 3 - 4 | 297 | 86 | |
AK311890.1 | - | 3 - 4 | 286 | 86 | |
BC005373.1 | - | 3 - 4 | 315 | 86 | |
BG773408.1 | - | 3 - 4 | 426 | 86 | |
BP369345.1 | - | 3 - 4 | 435 | 86 |