The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199165.2 | NP_001186094.1 | 3 - 4 | 435 | 72 |
NM_001302891.1 | NP_001289820.1 | 3 - 4 | 511 | 72 | |
XM_005257119.4 | XP_005257176.1 | 3 - 4 | 526 | 72 | |
XM_006721740.2 | XP_006721803.1 | 3 - 4 | 398 | 72 | |
XM_006721744.3 | XP_006721807.1 | 3 - 4 | 530 | 72 | |
XM_011524461.2 | XP_011522763.1 | 3 - 4 | 542 | 72 | |
XM_011524462.2 | XP_011522764.1 | 3 - 4 | 528 | 72 | |
XM_011524463.2 | XP_011522765.1 | 3 - 4 | 528 | 72 | |
XM_011524464.2 | XP_011522766.1 | 3 - 4 | 528 | 72 | |
XM_017024302.1 | XP_016879791.1 | 3 - 4 | 398 | 72 | |
XM_017024303.1 | XP_016879792.1 | 3 - 4 | 530 | 72 | |
GenBank mRNA | AK096929.1 | - | 3 - 4 | 398 | 72 |
AK299075.1 | - | 3 - 4 | 386 | 72 | |
BC037991.2 | - | 3 - 4 | 497 | 72 | |
BC131805.1 | - | 3 - 4 | 339 | 72 | |
DC327965.1 | - | 3 - 4 | 511 | 72 |