Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRK2
Entrez Gene ID
120892
Gene Name
leucine rich repeat kinase 2
Gene Aliases
AURA17, DARDARIN, PARK8, RIPK7, ROCO2
Chromosome Location
Chr.12: 40224895 - 40369285 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198578.3 NP_940980.3 48 - 49 7300 139
XM_005268629.3 XP_005268686.1 48 - 49 7416 139
XM_011537877.2 XP_011536179.1 48 - 49 7416 139
XM_017018787.1 XP_016874276.1 27 - 28 4244 139
XM_017018788.1 XP_016874277.1 23 - 24 3488 139
GenBank mRNA AK131537.1 - 23 - 24 3604 139
AL832453.2 - 27 - 28 139
AL834529.1 - 5 - 6 798 139
AY792511.1 - 48 - 49 7179 139
BC117180.1 - 48 - 49 139

View Full Product Details