The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003642.3 | NP_003633.1 | 7 - 8 | 792 | 105 |
NR_027862.1 | 6 - 7 | 716 | 105 | ||
XM_006712808.3 | XP_006712871.1 | 8 - 9 | 988 | 105 | |
XM_011512027.2 | XP_011510329.1 | 7 - 8 | 1544 | 105 | |
XM_017005136.1 | XP_016860625.1 | 8 - 9 | 1765 | 105 | |
XM_017005137.1 | XP_016860626.1 | 8 - 9 | 1697 | 105 | |
GenBank mRNA | AF030424.1 | - | 7 - 8 | 752 | 105 |
AK309001.1 | - | 6 - 7 | 634 | 105 | |
AK313699.1 | - | 7 - 8 | 783 | 105 | |
BC018682.1 | - | 3 - 4 | 268 | 105 | |
BC063003.1 | - | 6 - 7 | 693 | 105 |