The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270521.1 | NP_001257450.1 | 13 - 14 | 1552 | 162 |
NM_001270522.1 | NP_001257451.1 | 13 - 14 | 1552 | 162 | |
NM_004830.3 | NP_004821.2 | 13 - 14 | 1552 | 162 | |
NM_015979.3 | NP_057063.2 | 14 - 15 | 1570 | 162 | |
XM_005267223.2 | XP_005267280.1 | 14 - 15 | 1566 | 162 | |
XM_006715612.2 | XP_006715675.1 | 14 - 15 | 1566 | 162 | |
XM_011536257.2 | XP_011534559.1 | 12 - 13 | 1452 | 162 | |
XM_017011501.1 | XP_016866990.1 | 4 - 5 | 510 | 162 | |
GenBank mRNA | AB033042.1 | - | 13 - 14 | 1521 | 162 |
AF104255.1 | - | 14 - 15 | 1530 | 162 | |
AF105332.1 | - | 13 - 14 | 1481 | 162 | |
AK304711.1 | - | 2 - 3 | 291 | 162 | |
AK315030.1 | - | 13 - 14 | 1433 | 162 | |
AL136776.1 | - | 14 - 15 | 1536 | 162 | |
BC060759.1 | - | 13 - 14 | 1496 | 162 | |
EU392527.1 | - | 4 - 5 | 270 | 162 |