The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127208.2 | NP_001120680.1 | 8 - 9 | 4538 | 87 |
XM_005263082.2 | XP_005263139.1 | 7 - 8 | 4414 | 87 | |
XM_006714242.3 | XP_006714305.1 | 6 - 7 | 4082 | 87 | |
GenBank mRNA | AB046766.1 | - | 1 - 2 | 98 | 87 |
AB075496.1 | - | 5 - 6 | 1592 | 87 | |
BC110509.1 | - | 3 - 4 | 462 | 87 | |
BC110510.2 | - | 3 - 4 | 462 | 87 | |
FM992369.1 | - | 8 - 9 | 4911 | 87 |