The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184785.1 | NP_001171714.1 | 19 - 20 | 3161 | 109 |
NM_001184786.1 | NP_001171715.1 | 18 - 19 | 3122 | 109 | |
NM_001184787.1 | NP_001171716.1 | 19 - 20 | 3170 | 109 | |
NM_001184788.1 | NP_001171717.1 | 18 - 19 | 3032 | 109 | |
NM_001184789.1 | NP_001171718.1 | 18 - 19 | 3032 | 109 | |
NM_001184790.1 | NP_001171719.1 | 17 - 18 | 2900 | 109 | |
NM_001184791.1 | NP_001171720.1 | 16 - 17 | 2945 | 109 | |
NM_001184792.1 | NP_001171721.1 | 19 - 20 | 3161 | 109 | |
NM_001184793.1 | NP_001171722.1 | 19 - 20 | 3068 | 109 | |
NM_001184794.1 | NP_001171723.1 | 18 - 19 | 3032 | 109 | |
NM_019619.3 | NP_062565.2 | 19 - 20 | 3170 | 109 | |
XM_005252528.4 | XP_005252585.1 | 19 - 20 | 3149 | 109 | |
XM_005252530.4 | XP_005252587.1 | 18 - 19 | 3107 | 109 | |
XM_005252531.4 | XP_005252588.1 | 19 - 20 | 3062 | 109 | |
XM_005252532.4 | XP_005252589.1 | 18 - 19 | 2806 | 109 | |
XM_005252534.4 | XP_005252591.1 | 19 - 20 | 3149 | 109 | |
XM_005252535.4 | XP_005252592.1 | 18 - 19 | 3110 | 109 | |
XM_011519569.2 | XP_011517871.1 | 19 - 20 | 3158 | 109 | |
XM_011519570.2 | XP_011517872.1 | 18 - 19 | 3122 | 109 | |
XM_011519571.2 | XP_011517873.1 | 18 - 19 | 3116 | 109 | |
XM_011519572.2 | XP_011517874.1 | 18 - 19 | 3113 | 109 | |
XM_011519573.2 | XP_011517875.1 | 19 - 20 | 3071 | 109 | |
XM_011519575.2 | XP_011517877.1 | 18 - 19 | 2845 | 109 | |
XM_011519576.2 | XP_011517878.1 | 18 - 19 | 2816 | 109 | |
XM_011519577.2 | XP_011517879.1 | 17 - 18 | 2993 | 109 | |
XM_011519578.2 | XP_011517880.1 | 17 - 18 | 2991 | 109 | |
XM_011519579.2 | XP_011517881.1 | 17 - 18 | 2714 | 109 | |
XM_011519581.2 | XP_011517883.1 | 19 - 20 | 3161 | 109 | |
XM_011519583.2 | XP_011517885.1 | 19 - 20 | 3161 | 109 | |
XM_017016430.1 | XP_016871919.1 | 18 - 19 | 3104 | 109 | |
GenBank mRNA | AB073671.1 | - | 16 - 17 | 2615 | 109 |
AF177228.1 | - | 12 - 13 | 1816 | 109 | |
AF196185.1 | - | 19 - 20 | 3118 | 109 | |
AF196186.1 | - | 19 - 20 | 3118 | 109 | |
AF332592.1 | - | 18 - 19 | 2702 | 109 | |
AF332593.1 | - | 18 - 19 | 2702 | 109 | |
AF454057.1 | - | 17 - 18 | 2538 | 109 | |
AF454058.1 | - | 17 - 18 | 3010 | 109 | |
AF454059.1 | - | 14 - 15 | 2122 | 109 | |
AF467002.1 | - | 19 - 20 | 2840 | 109 | |
AF467003.1 | - | 19 - 20 | 2831 | 109 | |
AF467004.1 | - | 19 - 20 | 2840 | 109 | |
AF467005.1 | - | 16 - 17 | 2615 | 109 | |
AF467006.1 | - | 18 - 19 | 2702 | 109 | |
AK024668.1 | - | 6 - 7 | 813 | 109 | |
AK025892.1 | - | 10 - 11 | 1608 | 109 | |
AK027735.1 | - | 17 - 18 | 2365 | 109 | |
BC011711.2 | - | 2 - 3 | 263 | 109 | |
BC071566.1 | - | 18 - 19 | 3069 | 109 | |
BM796605.1 | - | 2 - 3 | 264 | 109 |