The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130852.1 | NP_001124324.1 | 9 - 10 | 1356 | 151 |
NM_012116.3 | NP_036248.3 | 10 - 11 | 1494 | 151 | |
GenBank mRNA | AB028645.1 | - | 10 - 11 | 1472 | 151 |
AF117646.1 | - | 10 - 11 | 1494 | 151 | |
AF117647.1 | - | 9 - 10 | 1356 | 151 | |
BC028915.1 | - | 10 - 11 | 1452 | 151 |