The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282745.1 | NP_001269674.1 | 14 - 15 | 1914 | 160 |
XM_017015831.1 | XP_016871320.1 | 13 - 14 | 1702 | 160 | |
XM_017015832.1 | XP_016871321.1 | 14 - 15 | 1897 | 160 | |
GenBank mRNA | AK057389.1 | - | 14 - 15 | 1857 | 160 |
AK302985.1 | - | 14 - 15 | 1901 | 160 |