The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100398.1 | NP_001093868.1 | 16 - 17 | 1540 | 105 |
NM_015085.4 | NP_055900.4 | 17 - 18 | 1585 | 105 | |
XM_005256542.2 | XP_005256599.1 | 16 - 17 | 1977 | 105 | |
XM_006721477.3 | XP_006721540.1 | 17 - 18 | 1526 | 105 | |
XM_011523738.1 | XP_011522040.1 | 18 - 19 | 1638 | 105 | |
XM_011523739.1 | XP_011522041.1 | 17 - 18 | 1593 | 105 | |
XM_011523740.2 | XP_011522042.1 | 17 - 18 | 1498 | 105 | |
XM_011523741.2 | XP_011522043.1 | 17 - 18 | 1674 | 105 | |
XM_011523742.2 | XP_011522044.1 | 14 - 15 | 1380 | 105 | |
XM_011523743.2 | XP_011522045.1 | 15 - 16 | 1458 | 105 | |
XM_011523744.2 | XP_011522046.1 | 10 - 11 | 1046 | 105 | |
XM_011523745.2 | XP_011522047.1 | 11 - 12 | 1133 | 105 | |
XM_017024370.1 | XP_016879859.1 | 17 - 18 | 1563 | 105 | |
XM_017024371.1 | XP_016879860.1 | 16 - 17 | 1616 | 105 | |
GenBank mRNA | AB028962.1 | - | 6 - 7 | 639 | 105 |
AJ628446.1 | - | 17 - 18 | 1585 | 105 | |
AJ628447.1 | - | 16 - 17 | 1540 | 105 | |
AK124640.1 | - | 17 - 18 | 1674 | 105 | |
BC140870.1 | - | 17 - 18 | 1541 | 105 | |
BC140871.1 | - | 17 - 18 | 1541 | 105 | |
CR627427.1 | - | 16 - 17 | 1509 | 105 |