The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100398.1 | NP_001093868.1 | 22 - 23 | 2230 | 78 |
NM_015085.4 | NP_055900.4 | 23 - 24 | 2275 | 78 | |
XM_005256542.2 | XP_005256599.1 | 22 - 23 | 2667 | 78 | |
XM_006721477.3 | XP_006721540.1 | 23 - 24 | 2216 | 78 | |
XM_011523738.1 | XP_011522040.1 | 24 - 25 | 2328 | 78 | |
XM_011523739.1 | XP_011522041.1 | 23 - 24 | 2283 | 78 | |
XM_011523740.2 | XP_011522042.1 | 23 - 24 | 2188 | 78 | |
XM_011523741.2 | XP_011522043.1 | 23 - 24 | 2364 | 78 | |
XM_011523742.2 | XP_011522044.1 | 20 - 21 | 2070 | 78 | |
XM_011523743.2 | XP_011522045.1 | 21 - 22 | 2148 | 78 | |
XM_011523744.2 | XP_011522046.1 | 16 - 17 | 1736 | 78 | |
XM_011523745.2 | XP_011522047.1 | 17 - 18 | 1823 | 78 | |
XM_017024370.1 | XP_016879859.1 | 23 - 24 | 2253 | 78 | |
XM_017024371.1 | XP_016879860.1 | 22 - 23 | 2306 | 78 | |
GenBank mRNA | AB028962.1 | - | 12 - 13 | 1329 | 78 |
AJ628446.1 | - | 23 - 24 | 2275 | 78 | |
AJ628447.1 | - | 22 - 23 | 2230 | 78 | |
AK124640.1 | - | 23 - 24 | 2364 | 78 | |
BC140870.1 | - | 23 - 24 | 2231 | 78 | |
BC140871.1 | - | 23 - 24 | 2231 | 78 | |
CR627427.1 | - | 22 - 23 | 78 | ||
DN831680.1 | - | 6 - 7 | 584 | 78 |