Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL17D
Entrez Gene ID
53342
Gene Name
interleukin 17D
Gene Aliases
IL-17D
Chromosome Location
Chr.13: 20702775 - 20723100 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005266421.3 XP_005266478.1 1 - 2 543 76

View Full Product Details