Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRIP2
Entrez Gene ID
83714
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.12: 2825348 - 2835536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031474.2 NP_113662.1 1 - 2 411 71
XM_005253797.3 XP_005253854.1 1 - 2 562 71
XM_005253798.3 XP_005253855.1 1 - 2 562 71
XM_011521031.2 XP_011519333.1 2 - 3 353 71
XM_017020000.1 XP_016875489.1 1 - 2 411 71
XM_017020001.1 XP_016875490.1 2 - 3 353 71
GenBank mRNA AK098063.1 - 1 - 2 382 71
AK300944.1 - 2 - 3 347 71
AL136557.1 - 1 - 2 379 71
BC036063.1 - 1 - 2 380 71
BC131562.1 - 1 - 2 368 71
DA391605.1 - 1 - 2 411 71

View Full Product Details