The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031474.2 | NP_113662.1 | 3 - 4 | 647 | 64 |
XM_005253797.3 | XP_005253854.1 | 3 - 4 | 798 | 64 | |
XM_005253798.3 | XP_005253855.1 | 3 - 4 | 798 | 64 | |
XM_011521031.2 | XP_011519333.1 | 4 - 5 | 589 | 64 | |
XM_017020000.1 | XP_016875489.1 | 3 - 4 | 647 | 64 | |
XM_017020001.1 | XP_016875490.1 | 4 - 5 | 589 | 64 | |
GenBank mRNA | AK098063.1 | - | 3 - 4 | 618 | 64 |
AK300944.1 | - | 4 - 5 | 583 | 64 | |
AL136557.1 | - | 3 - 4 | 615 | 64 | |
BC036063.1 | - | 3 - 4 | 616 | 64 | |
BC131562.1 | - | 3 - 4 | 604 | 64 |