The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322793.1 | NP_001309722.1 | 4 - 5 | 737 | 71 |
NM_001322795.1 | NP_001309724.1 | 5 - 6 | 919 | 71 | |
NM_001322796.1 | NP_001309725.1 | 4 - 5 | 901 | 71 | |
NM_004969.3 | NP_004960.2 | 4 - 5 | 737 | 71 | |
NR_136399.1 | 4 - 5 | 737 | 71 | ||
XM_017016187.1 | XP_016871676.1 | 4 - 5 | 660 | 71 | |
XM_017016188.1 | XP_016871677.1 | 4 - 5 | 1254 | 71 | |
XM_017016189.1 | XP_016871678.1 | 5 - 6 | 653 | 71 | |
XM_017016190.1 | XP_016871679.1 | 4 - 5 | 780 | 71 | |
GenBank mRNA | AK312810.1 | - | 4 - 5 | 662 | 71 |
BC096336.1 | - | 4 - 5 | 701 | 71 | |
BC096337.1 | - | 4 - 5 | 701 | 71 | |
BC096339.3 | - | 4 - 5 | 701 | 71 | |
BX648462.1 | - | 4 - 5 | 902 | 71 | |
M21188.1 | - | 4 - 5 | 718 | 71 |