Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A9
Entrez Gene ID
114571
Gene Name
solute carrier family 22 member 9
Gene Aliases
HOAT4, OAT4, OAT7, UST3H, ust3
Chromosome Location
Chr.11: 63368871 - 63410878 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080866.2 NP_543142.2 9 - 10 1873 70
XM_017017160.1 XP_016872649.1 8 - 9 1450 70
GenBank mRNA AB062418.1 - 9 - 10 1706 70
AB074812.1 - 9 - 10 1854 70
AF440402.1 - 9 - 10 1841 70
AJ295270.1 - 9 - 10 1665 70
BC126288.1 - 9 - 10 1755 70

View Full Product Details