The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005139.2 | NP_005130.1 | 5 - 6 | 615 | 122 |
GenBank mRNA | AK313945.1 | - | 5 - 6 | 390 | 122 |
BC000871.1 | - | 5 - 6 | 447 | 122 | |
BX336217.2 | - | 5 - 6 | 615 | 122 | |
CR407648.1 | - | 3 - 4 | 305 | 122 | |
M20560.1 | - | 4 - 5 | 351 | 122 | |
M63310.1 | - | 4 - 5 | 341 | 122 |