The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006392.3 | NP_006383.2 | 5 - 6 | 698 | 73 |
NR_027700.2 | 5 - 6 | 698 | 73 | ||
GenBank mRNA | AK292721.1 | - | 5 - 6 | 598 | 73 |
BC018421.1 | - | 4 - 5 | 558 | 73 | |
BC035369.1 | - | 5 - 6 | 588 | 73 | |
BC104791.1 | - | 5 - 6 | 698 | 73 | |
BC104793.1 | - | 5 - 6 | 698 | 73 | |
BX647439.1 | - | 5 - 6 | 598 | 73 | |
Y12065.1 | - | 4 - 5 | 608 | 73 |