The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012116.3 | NP_036248.3 | 5 - 6 | 980 | 91 |
XM_005258696.3 | XP_005258753.1 | 5 - 6 | 987 | 91 | |
XM_011526688.2 | XP_011524990.1 | 5 - 6 | 988 | 91 | |
GenBank mRNA | AB028645.1 | - | 5 - 6 | 958 | 91 |
AF117646.1 | - | 5 - 6 | 980 | 91 | |
BC028915.1 | - | 5 - 6 | 938 | 91 |