Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LINC01551
- Entrez Gene ID
-
387978
- Gene Name
- long intergenic non-protein coding RNA 1551
- Gene Aliases
-
C14orf23, c14_5148
- Chromosome Location
-
Chr.14: 28772704 - 28794794 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026731.1 |
|
3 - 3 |
385 |
69 |
GenBank mRNA |
BX248251.1 |
- |
3 - 3 |
292 |
69 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW160433.1 |
- |
3 |
273 |
|
Chr. 14 - 28772816 - 28792255 [+] on Build GRCh38 |
69 |
GenBankmRNA |
BX419086.2 |
- |
3 |
291 |
|
Chr. 14 - 28772796 - 28792615 [+] on Build GRCh38 |
69 |
GenBankmRNA |
DA254146.1 |
- |
1 |
194 |
|
Chr. 14 - 28791845 - 28792424 [+] on Build GRCh38 |
69 |
GenBankmRNA |
AI929489.1 |
- |
3 |
259 |
|
Chr. 14 - 28772830 - 28792246 [+] on Build GRCh38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105370424 |
LOC105370424 |
|
-6.1kb
|
Anti-sense |
Chr. - 28783491 - 28785787 on Build GRCh38 |
Downstream |
protein kinase D1 |
PRKD1 |
PKC-MU, PKCM, PKD, PRKCM |
784.2kb
|
Anti-sense |
Chr. - 29576479 - 29927693 on Build GRCh38 |
View Full Product Details