Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288814.1 NP_001275743.1 14 - 15 1568 78
NM_032450.2 NP_115826.2 15 - 16 1681 78
XM_011517270.2 XP_011515572.1 15 - 16 1819 78
XM_011517271.2 XP_011515573.1 15 - 16 1818 78
XM_011517272.2 XP_011515574.1 15 - 16 1819 78
XM_011517273.2 XP_011515575.1 15 - 16 1817 78
XM_011517274.2 XP_011515576.1 15 - 16 1817 78
XM_011517275.2 XP_011515577.1 15 - 16 1817 78
XM_011517276.2 XP_011515578.1 15 - 16 1845 78
XM_011517277.1 XP_011515579.1 15 - 16 1681 78
XM_011517278.1 XP_011515580.1 14 - 15 1561 78
XM_011517279.2 XP_011515581.1 14 - 15 1725 78
XM_011517281.1 XP_011515583.1 14 - 15 1734 78
XM_011517282.1 XP_011515584.1 13 - 14 1659 78
XM_011517283.2 XP_011515585.1 15 - 16 1640 78
XM_011517285.1 XP_011515587.1 14 - 15 1561 78
XM_017013802.1 XP_016869291.1 15 - 16 1817 78
XM_017013803.1 XP_016869292.1 15 - 16 1816 78
XM_017013804.1 XP_016869293.1 15 - 16 1681 78
XM_017013805.1 XP_016869294.1 14 - 15 1561 78
XM_017013806.1 XP_016869295.1 12 - 13 1296 78
GenBank mRNA AB058736.1 - 14 - 15 1573 78
AK092861.1 - 4 - 5 361 78
AK295740.1 - 14 - 15 1561 78
AL834309.1 - 14 - 15 1565 78

View Full Product Details