Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCRL4
Entrez Gene ID
83417
Gene Name
Fc receptor like 4
Gene Aliases
CD307d, FCRH4, IGFP2, IRTA1
Chromosome Location
Chr.1: 157573749 - 157598080 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031282.2 NP_112572.1 5 - 6 983 56
XM_011510034.1 XP_011508336.1 5 - 6 959 56
GenBank mRNA AF329490.1 - 5 - 6 847 56
AF329492.1 - 3 - 4 588 56
AF343659.1 - 5 - 6 983 56
AF343660.1 - 5 - 6 983 56
AF343661.1 - 5 - 6 983 56
AF397452.1 - 5 - 6 847 56
BC125173.1 - 5 - 6 941 56
BC125174.1 - 5 - 6 941 56
BC143628.1 - 4 - 5 655 56

View Full Product Details