The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014856.2 | NP_055671.2 | 21 - 22 | 3849 | 114 |
XM_005245678.4 | XP_005245735.1 | 21 - 22 | 4006 | 114 | |
XM_005245679.2 | XP_005245736.1 | 21 - 22 | 3856 | 114 | |
XM_011510219.1 | XP_011508521.1 | 19 - 20 | 3255 | 114 | |
XM_011510220.2 | XP_011508522.1 | 20 - 21 | 3669 | 114 | |
XM_011510221.2 | XP_011508523.1 | 19 - 20 | 3377 | 114 | |
XM_017002993.1 | XP_016858482.1 | 18 - 19 | 3231 | 114 | |
GenBank mRNA | AB007945.2 | - | 21 - 22 | 3849 | 114 |
AK226053.1 | - | 16 - 17 | 2504 | 114 |