The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014856.2 | NP_055671.2 | 2 - 3 | 715 | 88 |
XM_005245678.4 | XP_005245735.1 | 2 - 3 | 872 | 88 | |
XM_005245679.2 | XP_005245736.1 | 2 - 3 | 722 | 88 | |
XM_011510219.1 | XP_011508521.1 | 1 - 2 | 337 | 88 | |
XM_011510220.2 | XP_011508522.1 | 2 - 3 | 705 | 88 | |
XM_011510221.2 | XP_011508523.1 | 2 - 3 | 704 | 88 | |
XM_011510222.1 | XP_011508524.1 | 1 - 2 | 337 | 88 | |
XM_017002993.1 | XP_016858482.1 | 1 - 2 | 337 | 88 | |
GenBank mRNA | AB007945.2 | - | 2 - 3 | 715 | 88 |