The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014856.2 | NP_055671.2 | 7 - 8 | 1525 | 72 |
XM_005245678.4 | XP_005245735.1 | 7 - 8 | 1682 | 72 | |
XM_005245679.2 | XP_005245736.1 | 7 - 8 | 1532 | 72 | |
XM_011510219.1 | XP_011508521.1 | 6 - 7 | 1147 | 72 | |
XM_011510220.2 | XP_011508522.1 | 6 - 7 | 1345 | 72 | |
XM_011510222.1 | XP_011508524.1 | 6 - 7 | 1147 | 72 | |
XM_017002993.1 | XP_016858482.1 | 4 - 5 | 907 | 72 | |
GenBank mRNA | AB007945.2 | - | 7 - 8 | 1525 | 72 |
AK226053.1 | - | 2 - 3 | 174 | 72 |