Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
Chromosome Location
Chr.10: 71396934 - 71815947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171933.1 NP_001165404.1 3 - 4 649 115
NM_001171934.1 NP_001165405.1 3 - 4 649 115
NM_022124.5 NP_071407.4 50 - 51 7442 115
XM_006717940.3 XP_006718003.1 51 - 52 7496 115
XM_006717942.3 XP_006718005.1 49 - 50 7430 115
XM_011540039.2 XP_011538341.1 50 - 51 7493 115
XM_011540042.2 XP_011538344.1 52 - 53 7406 115
XM_011540043.2 XP_011538345.1 51 - 52 7496 115
XM_011540044.2 XP_011538346.1 51 - 52 7137 115
XM_011540045.2 XP_011538347.1 51 - 52 7496 115
XM_011540046.2 XP_011538348.1 46 - 47 6931 115
XM_011540047.2 XP_011538349.1 40 - 41 6231 115
XM_011540052.2 XP_011538354.1 23 - 24 3771 115
XM_017016499.1 XP_016871988.1 50 - 51 7442 115
XM_017016500.1 XP_016871989.1 50 - 51 7370 115
XM_017016501.1 XP_016871990.1 30 - 31 4998 115
XM_017016502.1 XP_016871991.1 30 - 31 5186 115
XM_017016503.1 XP_016871992.1 20 - 21 3174 115
XM_017016504.1 XP_016871993.1 14 - 15 2959 115
XM_017016505.1 XP_016871994.1 14 - 15 2990 115
XM_017016506.1 XP_016871995.1 13 - 14 2429 115
XM_017016507.1 XP_016871996.1 7 - 8 1251 115
XM_017016508.1 XP_016871997.1 3 - 4 704 115
GenBank mRNA AB053445.1 - 2 - 3 588 115
AF312024.1 - 49 - 50 7052 115
AY010111.2 - 49 - 50 7052 115
AY563165.1 - 3 - 4 649 115
AY563166.1 - 3 - 4 649 115

View Full Product Details