Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
Chromosome Location
Chr.10: 71396934 - 71815947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171933.1 NP_001165404.1 13 - 14 2320 56
NM_001171934.1 NP_001165405.1 13 - 14 2320 56
NM_022124.5 NP_071407.4 60 - 61 9113 56
XM_006717940.3 XP_006718003.1 61 - 62 9167 56
XM_006717942.3 XP_006718005.1 59 - 60 9101 56
XM_011540039.2 XP_011538341.1 60 - 61 9164 56
XM_011540042.2 XP_011538344.1 62 - 63 9077 56
XM_011540043.2 XP_011538345.1 61 - 62 9167 56
XM_011540044.2 XP_011538346.1 61 - 62 8808 56
XM_011540045.2 XP_011538347.1 61 - 62 9167 56
XM_011540046.2 XP_011538348.1 56 - 57 8602 56
XM_011540047.2 XP_011538349.1 50 - 51 7902 56
XM_011540052.2 XP_011538354.1 33 - 34 5442 56
XM_017016499.1 XP_016871988.1 60 - 61 9113 56
XM_017016500.1 XP_016871989.1 60 - 61 9041 56
XM_017016501.1 XP_016871990.1 40 - 41 6669 56
XM_017016502.1 XP_016871991.1 40 - 41 6857 56
XM_017016503.1 XP_016871992.1 30 - 31 4845 56
XM_017016504.1 XP_016871993.1 24 - 25 4630 56
XM_017016505.1 XP_016871994.1 24 - 25 4661 56
XM_017016506.1 XP_016871995.1 23 - 24 4100 56
XM_017016507.1 XP_016871996.1 17 - 18 2922 56
XM_017016508.1 XP_016871997.1 13 - 14 2375 56
GenBank mRNA AB053445.1 - 12 - 13 2259 56
AF312024.1 - 59 - 60 8723 56
AY010111.2 - 59 - 60 8723 56
AY563165.1 - 13 - 14 2320 56
AY563166.1 - 13 - 14 2320 56

View Full Product Details