Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CDH23
- Entrez Gene ID
-
64072
- Gene Name
- cadherin-related 23
- Gene Aliases
-
CDHR23, USH1D
- Chromosome Location
-
Chr.10: 71396934 - 71815947 on Build GRCh38
View Full Product Details