Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL834145.1 - 7 - 8 652 94

View Full Product Details