Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA3
Entrez Gene ID
21
Gene Name
ATP binding cassette subfamily A member 3
Gene Aliases
ABC-C, ABC3, EST111653, LBM180, SMDP3
Chromosome Location
Chr.16: 2275878 - 2340746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089.2 NP_001080.2 7 - 8 1329 91
GenBank mRNA AB070929.1 - 4 - 5 659 91
AB210044.1 - 7 - 8 1329 91
AK310864.1 - 7 - 8 1180 91
BC140895.1 - 4 - 5 654 91
BC146866.1 - 4 - 5 654 91
BC151840.1 - 7 - 8 1311 91
X97187.1 - 6 - 7 964 91

View Full Product Details