The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000742.3 | NP_000733.2 | 1 - 2 | 475 | 70 |
NM_001282455.1 | NP_001269384.1 | 1 - 2 | 475 | 70 | |
XM_005273397.2 | XP_005273454.1 | 1 - 2 | 399 | 70 | |
XM_011544389.1 | XP_011542691.1 | 1 - 2 | 475 | 70 | |
XM_017013003.1 | XP_016868492.1 | 1 - 2 | 452 | 70 | |
GenBank mRNA | AK314787.1 | - | 1 - 2 | 393 | 70 |
DA407151.1 | - | 1 - 2 | 475 | 70 | |
U62431.1 | - | 1 - 2 | 420 | 70 |