The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004119.2 | NP_004110.2 | 5 - 6 | 694 | 116 |
NR_130706.1 | 5 - 6 | 694 | 116 | ||
XM_011535015.2 | XP_011533317.1 | 5 - 6 | 693 | 116 | |
XM_011535017.2 | XP_011533319.1 | 5 - 6 | 596 | 116 | |
XM_011535018.2 | XP_011533320.1 | 2 - 3 | 204 | 116 | |
XM_017020486.1 | XP_016875975.1 | 5 - 6 | 689 | 116 | |
XM_017020487.1 | XP_016875976.1 | 6 - 7 | 767 | 116 | |
GenBank mRNA | BC036028.1 | - | 5 - 6 | 694 | 116 |
BC126350.1 | - | 5 - 6 | 678 | 116 | |
BC144039.1 | - | 5 - 6 | 679 | 116 | |
BC144040.1 | - | 5 - 6 | 679 | 116 | |
U02687.1 | - | 5 - 6 | 669 | 116 | |
Z26652.1 | - | 5 - 6 | 612 | 116 |