Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RPS6KC1
- Entrez Gene ID
-
26750
- Gene Name
- ribosomal protein S6 kinase C1
- Gene Aliases
-
RPK118, RSKL1, S6K-delta-1, S6PKh1, humS6PKh1
- Chromosome Location
-
Chr.1: 213051233 - 213378753 on Build GRCh38
View Full Product Details