Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCS1
Entrez Gene ID
23413
Gene Name
neuronal calcium sensor 1
Gene Aliases
FLUP, FREQ
Chromosome Location
Chr.9: 130172578 - 130237304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128826.1 NP_001122298.1 4 - 5 268 60
NM_014286.3 NP_055101.2 4 - 5 399 60
GenBank mRNA AF134479.1 - 4 - 5 380 60
AF186409.1 - 4 - 5 326 60
BC004856.2 - 4 - 5 399 60
BQ880305.1 - 4 - 5 268 60
DA092531.1 - 4 - 5 267 60
X84048.1 - 4 - 5 327 60

View Full Product Details