The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317926.1 | NP_001304855.1 | 9 - 10 | 1651 | 92 |
NM_013355.4 | NP_037487.2 | 9 - 10 | 1651 | 92 | |
XM_005251946.3 | XP_005252003.1 | 9 - 10 | 2206 | 92 | |
XM_006717080.2 | XP_006717143.1 | 9 - 10 | 1720 | 92 | |
XM_017014649.1 | XP_016870138.1 | 9 - 10 | 1722 | 92 | |
XM_017014650.1 | XP_016870139.1 | 9 - 10 | 1860 | 92 | |
GenBank mRNA | AB019692.1 | - | 9 - 10 | 1239 | 92 |
BC062558.1 | - | 9 - 10 | 1254 | 92 |