Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VMP1
Entrez Gene ID
81671
Gene Name
vacuole membrane protein 1
Gene Aliases
EPG3, TANGO5, TMEM49
Chromosome Location
Chr.17: 59707502 - 59840593 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB047551.1 - 3 - 4 305 126
AF214006.1 - 4 - 5 418 126
AK024969.1 - 4 - 5 428 126
AK025987.1 - 3 - 4 317 126
AK293664.1 - 4 - 5 358 126
AK301254.1 - 3 - 4 294 126
AL136711.1 - 4 - 5 438 126
AX775965.1 - 4 - 5 418 126
BC009758.2 - 4 - 5 429 126
CR533521.1 - 3 - 4 305 126

View Full Product Details