Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR5C1
Entrez Gene ID
392391
Gene Name
olfactory receptor family 5 subfamily C member 1
Gene Aliases
OR5C2P, OR9-31, OR9-F
Chromosome Location
Chr.9: 122788933 - 122789895 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001923.1 NP_001001923.1 1 - 1 856 68
GenBank mRNA BC136700.1 - 1 - 1 918 68
BC136701.1 - 1 - 1 918 68

View Full Product Details