The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294340.1 | NP_001281269.1 | 11 - 12 | 2062 | 103 |
NM_144604.3 | NP_653205.3 | 10 - 11 | 1990 | 103 | |
XM_011522863.1 | XP_011521165.1 | 9 - 10 | 1675 | 103 | |
XM_011522865.1 | XP_011521167.1 | 3 - 4 | 432 | 103 | |
GenBank mRNA | AK091849.1 | - | 5 - 6 | 817 | 103 |
AK225807.1 | - | 10 - 11 | 1960 | 103 | |
AK300254.1 | - | 11 - 12 | 2040 | 103 | |
AK302716.1 | - | 11 - 12 | 1436 | 103 | |
BC001584.1 | - | 7 - 8 | 1086 | 103 | |
BC050463.1 | - | 10 - 11 | 1937 | 103 |