Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H18
Entrez Gene ID
124245
Gene Name
zinc finger CCCH-type containing 18
Gene Aliases
NHN1
Chromosome Location
Chr.16: 88570381 - 88631964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001294340.1 NP_001281269.1 4 - 5 965 104
XM_011522863.1 XP_011521165.1 4 - 5 965 104
XM_017022934.1 XP_016878423.1 4 - 5 965 104
GenBank mRNA AK300254.1 - 4 - 5 943 104

View Full Product Details