The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206998.1 | NP_001193927.1 | 7 - 8 | 1014 | 104 |
NM_032173.3 | NP_115549.2 | 7 - 8 | 807 | 104 | |
XM_011530435.2 | XP_011528737.1 | 8 - 9 | 1003 | 104 | |
XM_011530436.2 | XP_011528738.1 | 9 - 10 | 1058 | 104 | |
XM_011530437.1 | XP_011528739.1 | 8 - 9 | 1728 | 104 | |
XM_011530438.2 | XP_011528740.1 | 8 - 9 | 1244 | 104 | |
XM_017028990.1 | XP_016884479.1 | 8 - 9 | 1499 | 104 | |
GenBank mRNA | AB051436.1 | - | 7 - 8 | 879 | 104 |
AK025710.1 | - | 2 - 3 | 201 | 104 | |
AK096397.1 | - | 7 - 8 | 807 | 104 | |
BC009252.2 | - | 2 - 3 | 138 | 105 | |
BC021570.1 | - | 2 - 3 | 166 | 104 | |
BC069019.1 | - | 2 - 3 | 137 | 104 | |
BC094857.1 | - | 2 - 3 | 137 | 104 | |
CR456397.1 | - | 7 - 8 | 787 | 104 |