Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GOLGB1
- Entrez Gene ID
-
2804
- Gene Name
- golgin B1
- Gene Aliases
-
GCP, GCP372, GOLIM1
- Chromosome Location
-
Chr.3: 121663199 - 121749908 on Build GRCh38
View Full Product Details