The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017554.2 | NP_060024.2 | 9 - 10 | 3680 | 108 |
XM_011512928.2 | XP_011511230.1 | 9 - 10 | 3771 | 108 | |
GenBank mRNA | AB033094.1 | - | 4 - 5 | 1650 | 108 |
AK001770.1 | - | 5 - 6 | 708 | 108 | |
AK026003.1 | - | 4 - 5 | 1509 | 108 | |
AK304269.1 | - | 9 - 10 | 3648 | 108 | |
AY134858.1 | - | 4 - 5 | 2853 | 108 | |
BX648758.1 | - | 5 - 6 | 2999 | 108 | |
DQ063584.1 | - | 6 - 7 | 3144 | 108 | |
DQ063585.1 | - | 6 - 7 | 3144 | 108 |