Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
Chromosome Location
Chr.1: 91949371 - 92014428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001726.4 NP_001717.3 1 - 2 112
GenBank mRNA AF019085.1 - 1 - 2 112
BG717291.1 - 1 - 2 112
DC401352.1 - 1 - 2 107 113

View Full Product Details