The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261408.1 | NP_001248337.1 | 6 - 7 | 772 | 67 |
NM_001261409.1 | NP_001248338.1 | 4 - 5 | 758 | 67 | |
NM_001261410.1 | NP_001248339.1 | 2 - 3 | 425 | 67 | |
NM_003567.3 | NP_003558.1 | 4 - 5 | 735 | 67 | |
XM_017002480.1 | XP_016857969.1 | 6 - 7 | 1148 | 67 | |
GenBank mRNA | AF124250.1 | - | 4 - 5 | 727 | 67 |
AK055442.1 | - | 4 - 5 | 758 | 67 | |
AL833121.1 | - | 2 - 3 | 378 | 67 | |
BC028477.1 | - | 6 - 7 | 801 | 67 | |
BC039895.1 | - | 4 - 5 | 628 | 67 | |
DA057195.1 | - | 2 - 3 | 425 | 67 | |
U92715.1 | - | 4 - 5 | 543 | 67 |