Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAR3
Entrez Gene ID
8412
Gene Name
breast cancer anti-estrogen resistance 3
Gene Aliases
NSP2, SH2D3B
Chromosome Location
Chr.1: 93561741 - 93847150 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261408.1 NP_001248337.1 7 - 8 1215 66
NM_001261409.1 NP_001248338.1 5 - 6 1201 66
NM_001261410.1 NP_001248339.1 3 - 4 868 66
NM_003567.3 NP_003558.1 5 - 6 1178 66
XM_011542251.2 XP_011540553.1 4 - 5 817 66
XM_017002480.1 XP_016857969.1 7 - 8 1591 66
GenBank mRNA AF124250.1 - 5 - 6 1170 66
AK027785.1 - 1 - 2 436 66
AK055442.1 - 5 - 6 1201 66
AL833121.1 - 3 - 4 821 66
BC028477.1 - 7 - 8 1244 66
BC039895.1 - 5 - 6 1071 66
DA745974.1 - 1 - 2 151 66
U92715.1 - 5 - 6 986 66

View Full Product Details