Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAR3
Entrez Gene ID
8412
Gene Name
breast cancer anti-estrogen resistance 3
Gene Aliases
NSP2, SH2D3B
Chromosome Location
Chr.1: 93561741 - 93847150 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261408.1 NP_001248337.1 1 - 2 83 88
GenBank mRNA BC028477.1 - 1 - 2 112 88

View Full Product Details