The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000773.3 | NP_000764.1 | 2 - 3 | 364 | 68 |
GenBank mRNA | AF084225.1 | - | 2 - 3 | 238 | 68 |
AF182276.1 | - | 2 - 3 | 339 | 68 | |
AK290822.1 | - | 2 - 3 | 364 | 68 | |
BC067433.1 | - | 2 - 3 | 334 | 68 | |
BX951478.1 | - | 2 - 3 | 365 | 68 | |
DQ149222.1 | - | 2 - 3 | 348 | 68 | |
J02625.1 | - | 2 - 3 | 343 | 68 |