The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004947.4 | NP_004938.1 | 16 - 17 | 1561 | 78 |
XM_005264914.3 | XP_005264971.1 | 16 - 17 | 1833 | 78 | |
XM_005264915.3 | XP_005264972.1 | 16 - 17 | 1833 | 78 | |
XM_005264916.3 | XP_005264973.1 | 16 - 17 | 1611 | 78 | |
XM_005264917.3 | XP_005264974.1 | 16 - 17 | 1833 | 78 | |
XM_005264918.3 | XP_005264975.1 | 16 - 17 | 1610 | 78 | |
XM_006713008.3 | XP_006713071.1 | 16 - 17 | 1833 | 78 | |
XM_006713009.3 | XP_006713072.1 | 16 - 17 | 1833 | 78 | |
XM_006713010.3 | XP_006713073.1 | 16 - 17 | 1596 | 78 | |
XM_011533441.2 | XP_011531743.1 | 5 - 6 | 2442 | 78 | |
XM_017005825.1 | XP_016861314.1 | 16 - 17 | 1612 | 78 | |
XM_017005826.1 | XP_016861315.1 | 16 - 17 | 1602 | 78 | |
XM_017005827.1 | XP_016861316.1 | 7 - 8 | 753 | 78 | |
GenBank mRNA | AB002297.1 | - | 11 - 12 | 1169 | 78 |
AY145303.1 | - | 16 - 17 | 1538 | 78 | |
AY254099.1 | - | 16 - 17 | 1561 | 78 |