The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004947.4 | NP_004938.1 | 32 - 33 | 3426 | 95 |
XM_005264914.3 | XP_005264971.1 | 32 - 33 | 3698 | 95 | |
XM_005264915.3 | XP_005264972.1 | 32 - 33 | 3698 | 95 | |
XM_005264916.3 | XP_005264973.1 | 32 - 33 | 3476 | 95 | |
XM_005264917.3 | XP_005264974.1 | 32 - 33 | 3656 | 95 | |
XM_005264918.3 | XP_005264975.1 | 31 - 32 | 3328 | 95 | |
XM_006713008.3 | XP_006713071.1 | 32 - 33 | 3698 | 95 | |
XM_006713009.3 | XP_006713072.1 | 32 - 33 | 3698 | 95 | |
XM_006713010.3 | XP_006713073.1 | 32 - 33 | 3461 | 95 | |
XM_011533441.2 | XP_011531743.1 | 21 - 22 | 4307 | 95 | |
XM_011533443.2 | XP_011531745.1 | 8 - 9 | 1988 | 95 | |
XM_011533444.2 | XP_011531746.1 | 8 - 9 | 1912 | 95 | |
XM_011533445.2 | XP_011531747.1 | 8 - 9 | 1933 | 95 | |
XM_017005825.1 | XP_016861314.1 | 32 - 33 | 3477 | 95 | |
XM_017005826.1 | XP_016861315.1 | 32 - 33 | 3467 | 95 | |
XM_017005827.1 | XP_016861316.1 | 23 - 24 | 2618 | 95 | |
XM_017005828.1 | XP_016861317.1 | 7 - 8 | 1343 | 95 | |
GenBank mRNA | AB002297.1 | - | 27 - 28 | 3034 | 95 |
AY145303.1 | - | 32 - 33 | 3403 | 95 | |
AY254099.1 | - | 32 - 33 | 3426 | 95 |